EMAIL THIS PAGE TO A FRIEND
Cellular signalling
Store-operated calcium entry is dispensable for the activation of ERK1/2 pathway in prostate cancer cells.
PMID 28866365
Abstract
STIM1, the endoplasmic reticulum Ca