EMAIL THIS PAGE TO A FRIEND
Food technology and biotechnology
Genetic, Biochemical, Nutritional and Antimicrobial Characteristics of Pomegranate (
PMID 28867945
Abstract
This study characterises the genetic variability of local pomegranate (