EMAIL THIS PAGE TO A FRIEND
Food technology and biotechnology
Extracellular Alkaline Lipase from a Novel Fungus
PMID 28867950
Abstract
Thirty isolated fungal strains were screened for lipase production using Phenol Red plates, containing tributyrin as lipidic substrate, and a novel fungus identified genetically as
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W222305
Tributyrin, 97%, FG
C15H26O6