EMAIL THIS PAGE TO A FRIEND
Journal of geriatric cardiology : JGC
Glycation of high-density lipoprotein triggers oxidative stress and promotes the proliferation and migration of vascular smooth muscle cells.
PMID 28868076
Abstract
In type 2 diabetes mellitus (T2DM), high-density lipoprotein (HDL) impairs its anti-atherogenic properties and even develops to a pro-inflammatory and pro-atherogenic phenotype because of abnormal compositions and modifications. In this study, we examined the effects and the related mechanisms of glycation of HDL on the proliferation and migration of vascular smooth muscle cells (VSMCs). Glycated HDL (G-HDL) was modified with D-glucose (25 mmol/L) HDL either glycated