EMAIL THIS PAGE TO A FRIEND
Experimental cell research
Interleukin 10 enhanced CD8+ T cell activity and reduced CD8
PMID 28870600
Abstract
The pleiotropic cytokine interleukin (IL)-10 is best characterized by its ability to downregulate inflammation and promote peripheral tolerance. On the other hand, IL-10 was also found to maintain the effector response of CD8
Related Materials