EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
Engineering PLGA nano-based systems through understanding the influence of nanoparticle properties and cell-penetrating peptides for cochlear drug delivery.
PMID 28870763
Abstract
The properties of poly(lactic-co-glycolic acid) (PLGA) nanoparticles (NPs) and penetration enhancers play a deciding role in the inner ear drug delivery of NPs across the round window membrane (RWM). Thus, PLGA nano-based systems with a variety of particle sizes and surface chemistries and those combined with cell-penetrating peptides (CPPs) as penetration enhancers were devised to explore their impact on the cochlear drug delivery in vivo. First, we demonstrated that the properties of NPs dictated the extent of NP cochlear entry by near-infrared fluorescence imaging. NPs with the sizes of 150 and 300nm had faster entry than that of 80nm NPs. At 0.5h, among the NPs unmodified and modified with chitosan (CS), poloxamer 407 (P407) and methoxy polyethylene glycol, CS-PLGA-NPs (positive surface charge) carried payload to the cochlea fastest, whereas P407-PLGA-NPs (surface hydrophilicity) showed the greatest distribution in the cochlea at 24h. Compared to other CPPs (TAT, penetratin and poly(arginine)