EMAIL THIS PAGE TO A FRIEND
Scientific reports
Acupuncture and related therapies used as add-on or alternative to prokinetics for functional dyspepsia: overview of systematic reviews and network meta-analysis.
PMID 28871092
Abstract
Prokinetics for functional dyspepsia (FD) have relatively higher number needed to treat values. Acupuncture and related therapies could be used as add-on or alternative. An overview of systematic reviews (SRs) and network meta-analyses (NMA) were performed to evaluate the comparative effectiveness of different acupuncture and related therapies. We conducted a comprehensive literature search for SRs of randomized controlled trials (RCTs) in eight international and Chinese databases. Data from eligible RCTs were extracted for random effect pairwise meta-analyses. NMA was used to explore the most effective treatment among acupuncture and related therapies used alone or as add-on to prokinetics, compared to prokinetics alone. From five SRs, 22 RCTs assessing various acupuncture and related therapies were included. No serious adverse events were reported. Two pairwise meta-analyses showed manual acupuncture has marginally stronger effect in alleviating global FD symptoms, compared to domperidone or itopride. Results from NMA showed combination of manual acupuncture and clebopride has the highest probability in alleviating patient reported global FD symptom. Combination of manual acupuncture and clebopride has the highest probability of being the most effective treatment for FD symptoms. Patients who are contraindicated for prokinetics may use manual acupuncture or moxibustion as alternative. Future confirmatory comparative effectiveness trials should compare clebopride add-on manual acupuncture with domperidone add-on manual acupuncture and moxibustion.