EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Archaeal Persisters: Persister Cell Formation as a Stress Response in
PMID 28871247
Abstract
Persister cells are phenotypic variants within a microbial population, which are dormant and transiently tolerant to stress. Persistence has been studied extensively in bacteria, and in eukaryotes to a limited extent, however, it has never been observed in archaea. Using the model haloarchaeon,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5821
DS2, ≥98% (HPLC)
C18H12ClN3OS