EMAIL THIS PAGE TO A FRIEND
PMID 28874393
Abstract
Aerobic glycolysis plays an important role in cancer progression. New target genes regulating cancer aerobic glycolysis must be explored to improve patient prognosis. Mitochondrial topoisomerase I (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2751
Sodium oxamate, ≥98%
C2H2NO3Na