EMAIL THIS PAGE TO A FRIEND
Scientific reports
Mitochondrial permeability transition pore: sensitivity to opening and mechanistic dependence on substrate availability.
PMID 28874733
Abstract
Mitochondrial Ca