EMAIL THIS PAGE TO A FRIEND
Reproduction (Cambridge, England)
Histone 3 lysine 9 acetylation is a biomarker of the effects of culture on zygotes.
PMID 28878090
Abstract
Acetylation of histone proteins is a major determinant of chromatin structure and function. Fertilisation triggers a round of chromatin remodelling that prepares the genome for the first round of transcription from the new embryonic genome. In this study we confirm that fertilisation leads to a marked progressive increase in the level of histone 3 lysine 9 acetylation in both the paternally and maternally derived genomes. The culture of zygotes in simple defined media caused a marked increase in the global level of acetylation and this affected the male pronucleus more than the female. The culture created a marked asymmetry in staining between the two pronuclei that was not readily detected in zygotes collected directly from the reproductive tract and was ameliorated to some extent by optimized culture media. The increased acetylation caused by culture resulted in increased transcription of
Related Materials