EMAIL THIS PAGE TO A FRIEND
Physiology and molecular biology of plants : an international journal of functional plant biology
Comparative proteomic analysis of autotetraploid and diploid
PMID 28878499
Abstract
To enlarge the germplasm resource of