EMAIL THIS PAGE TO A FRIEND
Physiology and molecular biology of plants : an international journal of functional plant biology
An improved micropropagation system, ex vitro rooting and validation of genetic homogeneity in wild female
PMID 28878509
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9539
Agarose, BioReagent, for molecular biology, low EEO