EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Knockdown of SIRT7 enhances the osteogenic differentiation of human bone marrow mesenchymal stem cells partly via activation of the Wnt/β-catenin signaling pathway.
PMID 28880264
Abstract
Sirtuin 7 (SIRT7) is a NAD