EMAIL THIS PAGE TO A FRIEND
Oncotarget
Toxicarioside O induces protective autophagy in a sirtuin-1-dependent manner in colorectal cancer cells.
PMID 28881770
Abstract
Colorectal cancer is the most common cancer. It has high morbidity and mortality worldwide, and more effective treatment strategies need to be developed. Toxicarioside O (TCO), a natural product derived from
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7034
EX-527, ≥98% (HPLC)
C13H13ClN2O