EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Histone deacetylase 6 inhibition reduces cysts by decreasing cAMP and Ca
PMID 28887310
Abstract
Autosomal dominant polycystic kidney disease (ADPKD) is associated with progressive enlargement of multiple renal cysts, often leading to renal failure that cannot be prevented by a current treatment. Two proteins encoded by two genes are associated with ADPKD: PC1 (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU054851 MISSION® esiRNA, esiRNA human HDAC6 (esiRNA1)
SML0717
W-13, ≥98% (HPLC)
C14H17ClN2O2S · HCl