EMAIL THIS PAGE TO A FRIEND
Cancer research
Exercise-Induced Catecholamines Activate the Hippo Tumor Suppressor Pathway to Reduce Risks of Breast Cancer Development.
PMID 28887324
Abstract
Strong epidemiologic evidence documents the protective effect of physical activity on breast cancer risk, recurrence, and mortality, but the underlying mechanisms remain to be identified. Using human exercise-conditioned serum for breast cancer cell incubation studies and murine exercise interventions, we aimed to identify exercise factors and signaling pathways involved in the exercise-dependent suppression of breast cancer. Exercise-conditioned serum from both women with breast cancer (