EMAIL THIS PAGE TO A FRIEND
Genes & development
Enzymatic modules of the SAGA chromatin-modifying complex play distinct roles in
PMID 28887412
Abstract
The Spt-Ada-Gcn5-acetyltransferase (SAGA) chromatin-modifying complex is a transcriptional coactivator that contains four different modules of subunits. The intact SAGA complex has been well characterized for its function in transcription regulation and development. However, little is known about the roles of individual modules within SAGA and whether they have any SAGA-independent functions. Here we demonstrate that the two enzymatic modules of