EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Neuro-differentiated Ntera2 cancer stem cells encapsulated in alginate beads: First evidence of biological functionality.
PMID 28887978
Abstract
The present communication investigates an application of alginate encapsulation technology to the differentiation of the embryonic cancer stem NTera2 cells (NT2) into dopamine-producing cells. The encapsulation of cells in polymeric beads allows their immune isolation and makes them eligible for transplantation, thus representing a promising biotech tool for the delivery of biologically active compounds to the brain. The polysaccharide alginate is one of the most commonly used material for this procedure since it is well tolerated by various tissues, including the brain. Two different initial cell concentrations (i.e. 0.5∗10