EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
A targeted nanocarrier based on polyspermine for the effective delivery of methotrexate in nasopharyngeal carcinoma.
PMID 28888001
Abstract
Antitumor efficacy of methotrexate (MTX) is seriously limited due to its poor water solubility, nonspecific tumor distribution and low bioavailability. To overcome these obstacles, polyspermine (PSP) conjugated with MTX and folic acid (FA) as a novel targeted prodrug was designed and has been successfully synthesized using the amidation reaction. The strong hydrophilic properties of PSP made MTX well dispersed in water and the cellular uptake study indicated that the presence of FA enhanced uptake of the FA-PSP-MTX in folate receptor (FR) over-expressing human nasopharyngeal carcinoma HNE-1 cells.