EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Effects of ultraviolet light emitting diodes (LEDs) on microbial and enzyme inactivation of apple juice.
PMID 28888104
Abstract
In this study, the effects of Ultraviolet light-emitting diodes (UV-LEDs) on the inactivation of E. coli K12 (ATCC 25253), an indicator organism of E. coli O157:H7, and polyphneoloxidase (PPO) in cloudy apple juice (CAJ) were investigated. The clear (AJ) and cloudy apple juice were exposed to UV rays for 40min by using a UV device composed of four UV-LEDs with peak emissions at 254 and 280nm and coupled emissions as follows: 254/365, 254/405, 280/365, 280/405 and 254/280/365/405nm. UV-LEDs at 254nm achieved 1.6±0.1 log