EMAIL THIS PAGE TO A FRIEND
Environmental research
Does a lag-structure of temperature confound air pollution-lag-response relation? Simulation and application in 7 major cities, Korea (1998-2013).
PMID 28888197
Abstract
Temperature must be controlled when estimating the associations of short-term exposure to air pollution and mortality. Given that multi-country studies have implied temperature has lagged effects, we aim to explore confounding by temperature-lag-response and investigate PM In a simulation study, we compared the performance of different methods to control for: the same day temperature, a lagged temperature and distributed lags of temperature. In a real data study, we explored PM We confirmed that a model with insufficient control of temperature offers a biased estimate of PM A lag structure of temperature can confound the air pollution-lag-response relation. Temperature-lag-response relation should be evaluated when estimating air pollution-lag-response relation. As a corollary, air pollution and temperature risk in mortality can be estimated using the same regression model.