EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Quantitation of lanosterol in the vitreous humor of rabbits after ocular administration of lanosterol/thermogel formulation by ultra high performance liquid chromatography-tandem mass spectrometry with the electrospray ionization mode.
PMID 28888677
Abstract
Cataracts are the most common cause of blindness worldwide affecting tens of millions of people. Here, we report a simple, rapid, sensitive and specific method by ultra performance liquid chromatography-tandem mass spectrometry with the electrospray ionization mode (UPLC-ESI-MS/MS) for quantitation of lanosterol, a possible effective drug for cataracts, in the vitreous humor of rabbits after ocular administration. The injected lanosterol was prepared by dispersing lanosterol molecules into the poly-(dl-lactic acid-co-glycolic acid)-poly(ethylene glycol)-poly-(dl-lactic acid-co-glycolic acid) (PLGA-PEG-PLGA) thermogel solution. The analyte and internal standard (IS, panaxadiol) were extracted by the simple protein precipitation with methanol. The chromatographic separation used an Agilent RRHD SB-C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5768
Lanosterol, ≥93%, powder
C30H50O