EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Decontamination of fresh-cut cucumber slices by a combination of a modified chitosan coating containing carvacrol nanoemulsions and pulsed light.
PMID 28889001
Abstract
In this study, the impact of the combination of pulsed light (PL) treatments with antimicrobial coatings, consisting of modified chitosan suspensions incorporating carvacrol nanoemulsions, was investigated on the decontamination of fresh-cut cucumber slices. The upper surface of the cucumber slices, with or without the coating deposition, was inoculated with Escherichia coli ATCC 26 to reach a final concentration of 10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0400007 Glycerol mono-oleate, European Pharmacopoeia (EP) Reference Standard
C21H40O4