EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Prediction of arterial blood gas values from venous blood gas values in Asiatic black bears (Ursus thibetanus) anesthetized with intramuscular medetomidine and zolazepam-tiletamine.
PMID 28890467
Abstract
The objective of this study was to measure differences between arterial and venous blood gas parameters and to evaluate whether arterial blood gas values can be estimated from venous blood in Asiatic black bears (ABBs). Twelve healthy captive ABBs (8 males and 4 females; 8-16 years; 76.8-220 kg) were included in this study. The bears were immobilized with medetomidine and zolazepam-tiletamine using a dart gun. Arterial and venous samples were collected simultaneously at 5 and 35 min after recumbency (5- and 35-min points). Partial pressure of oxygen (PO