EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Identification of Fungal Communities Associated with the Biodeterioration of Waterlogged Archeological Wood in a Han Dynasty Tomb in China.
PMID 28890715
Abstract
The Mausoleum of the Dingtao King (termed 'M2') is a large-scale