Reproductive sciences (Thousand Oaks, Calif.)
Androgens Upregulate Endometrial Epithelial Progesterone Receptor Expression: Potential Implications for Endometriosis.
PMID 28891417
Androgenic compounds have been implicated in induction of endometrial atrophy yet the mechanisms of androgen effects on human endometrium have not been well studied. We hypothesized that androgens may promote their endometrial effects via modulation of progesterone receptor (PR) expression. Proliferative phase endometrial samples were collected at the time of hysterectomy. We evaluated the effect of the potent androgen 5α-dihydrotestosterone (DHT) on endometrial PR expression by treating human endometrial explants, endometrial stromal cells, and Ishikawa cells with DHT. Ishikawa cells were also treated with DHT ± the androgen receptor (AR) blocker flutamide. The PR-B, total PR messenger RNA (mRNA), and PR protein expression were assessed. Expression of cyclin D1 and D2 was checked as markers of cell proliferation. As expected, estradiol induced PR expression in isolated stromal cells, endometrial epithelial cells, and tissue explants. The DHT treatment also resulted in increased PR expression in endometrial explants and Ishikawa cells but not in stromal cells. Further, protein levels of both nuclear PR isoforms (PR-A and PR-B) were induced with the DHT treatment. Although flutamide treatment alone did not affect PR expression, flutamide diminished androgen-induced upregulation of PR in both endometrial explants and Ishikawa cells. Although estradiol induced both cyclin D1 and cyclin D2 mRNA, DHT did not induce these markers of cell proliferation. Androgens may mediate endometrial effects through upregulation of PR gene and protein expression. Endometrial PR upregulation by androgens is mediated, at least in part, through AR.