EMAIL THIS PAGE TO A FRIEND
The Analyst
Online monitoring of hepatic rat metabolism by coupling a liver biochip and a mass spectrometer.
PMID 28891561
Abstract
A microfluidic liver biochip was coupled with a mass spectrometer to detect in real time the drug metabolism of hepatocytes. The hepatocytes were cultivated in the biochip for 35 h. The biochip was placed in a small-scale incubator in which the temperature and CO
Related Materials