EMAIL THIS PAGE TO A FRIEND
Nature methods
CRISPR-Cas9-based photoactivatable transcription systems to induce neuronal differentiation.
PMID 28892089
Abstract
Our improved CRISPR-Cas9-based photoactivatable transcription systems, CPTS2.0 and Split-CPTS2.0, enable high blue-light-inducible activation of endogenous target genes in various human cell lines. We achieved reversible activation of target genes with CPTS2.0 and induced neuronal differentiation in induced pluripotent stem cells (iPSCs) by upregulating NEUROD1 with Split-CPTS2.0.