EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Bonding to CAD-CAM Composites: An Interfacial Fracture Toughness Approach.
PMID 28892655
Abstract
The objective of this study was to evaluate the interfacial fracture toughness (IFT) of composite cement with dispersed filler (DF) versus polymer-infiltrated ceramic network (PICN) computer-aided design and computer-aided manufacturing (CAD-CAM) composite blocks after 2 different surface pretreatments using the notchless triangular prism (NTP) test. Two DFs (Cerasmart [CRT] and Lava Ultimate [LVA]), 2 PICNs (Enamic [ENA] and experimental PICN [EXP]), and e.max CAD lithium disilicate glass-ceramic (EMX, control) prism samples were bonded to their counterparts with Variolink Esthetic DC composite cement after either hydrofluoric acid etching (HF) or gritblasting (GR). Both procedures were followed by silanization. All samples ( n = 30 per group) were thermocycled (10,000 cycles) and tested for their IFT in a water bath at 36°C. Moreover, representative samples from each group were subjected to a developed interfacial area ratio (Sdr) measurement by profilometry and scanning electron microscopy (SEM) characterization. EXP-HF gave the highest IFT (1.85 ± 0.39 MPa·m