EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Insight into the effects of modifying chromophores on the performance of quinoline-based dye-sensitized solar cells.
PMID 28892718
Abstract
A series of organic dyes based on quinoline as an electron-deficient π-linker, were designed and synthesized for dye sensitized solar cells (DSSC) application. These push-pull conjugated dyes, sharing same anchoring group with distinctive electron-rich donating groups such as N,N-diethyl (DEA-Q), 3,6-dimethoxy carbazole (CBZ-Q), bis(4-butoxyphenyl)amine (BPA-Q), were synthesized by Riley oxidation of CH