EMAIL THIS PAGE TO A FRIEND
BMC microbiology
Annexin A2 (ANXA2) interacts with nonstructural protein 1 and promotes the replication of highly pathogenic H5N1 avian influenza virus.
PMID 28893180
Abstract
Non-structural protein 1 (NS1) is a multifunctional protein and a crucial regulatory factor in the replication and pathogenesis of avian influenza virus (AIV). Studies have shown that NS1 can interact with a variety of host proteins to modulate the viral life cycle. We previously generated a monoclonal antibody against NS1 protein; In the current research study, using this antibody, we immunoprecipitated host proteins that interact with NS1 to better understand the roles played by NS1 in communications between virus and host. Co-immunoprecipitation experiments identified annexin A2 (ANXA2) as a target molecule interacting with NS1. Results from confocal laser scanning microscopy indicated that NS1 co-localized with ANXA2 in the cell cytoplasm. Overexpression of ANXA2 significantly increased the titer of H5N1 subtype HPAIV, whereas siRNA-mediated knockdown of ANXA2 markedly inhibited the expression of viral proteins and reduced the progeny virus titer. Our results indicate that ANXA2 interacts with NS1 and ANXA2 expression increases HPAIV replication.