EMAIL THIS PAGE TO A FRIEND
Scientific reports
Characterization of Volume-Based Changes in Cortical Auditory Evoked Potentials and Prepulse Inhibition.
PMID 28894145
Abstract
The auditory evoked startle reflex is a conserved response resulting in neurological and motor activity. The presence of a mild prepulse immediately before the main pulse inhibits startle responses, though the mechanism for this remains unknown. In this study, the electroencephalography (EEG) data recorded from 15 subjects was analyzed to study the N1 and P2 components of cortical auditory evoked potentials (CAEPs) evoked by 70, 80, 90, 100, and 110 dB stimuli both in the presence and absence of 70 dB prepulses. Results without a prepulse showed an evolution of N1 amplitudes, increasing with stimulus intensity and showing largely significant differences. Results from prepulse trials only showed noteworthy changes in peak-to-peak amplitude in the 100 dB condition. Prepulse and non-prepulse conditions were then compared using peak amplitudes and theta power. Prepulse conditions significantly decreased the amplitude for both components in the 110 dB condition, i.e., pre-pulse inhibition, but significantly increased the N1 amplitude in the 70 dB condition, i.e., pre-pulse facilitation. Similarly theta band power significantly increased in the 70 dB prepulse condition and significantly decreased in the 110 dB prepulse condition. These results expand the basis of knowledge regarding how CAEPs change and elaborate on their neural function and representation.