Scientific reports
Positive feedback of the amphiregulin-EGFR-ERK pathway mediates PM2.5 from wood smoke-induced MUC5AC expression in epithelial cells.
PMID 28894207
Biomass fuel smoke is thought to contribute to chronic obstructive pulmonary disease, which is characterized by mucous cell metaplasia and enhanced mucus secretion. We investigated the effect of particulate matter (PM) with a diameter <2.5 μm (PM2.5) from wood smoke (WSPM2.5) on the expression of the most prominent secreted mucin, MUC5AC. Wood smoke was able to induce MUC5AC expression in the rat respiratory tract after 3 months of exposure. WSPM2.5 could induce MUC5AC production in both primary human airway epithelial cells and the NCI-H292 cell line. This induction process was mediated by activation of epithelial growth factor receptor (EGFR)-extracellular signal-regulated kinase (ERK) signaling through an EGFR ligand-dependent mechanism. Amphiregulin (AR) was identified as the major ligand responsible for EGFR-ERK signaling activation and MUC5AC expression. In turn, EGFR-ERK pathway activation was found to contribute to the de novo synthesis of AR. This positive feedback loop might play an important role in a sustained mucus hypersecretion response.
Related Materials