EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Downregulation of UBE2E2 in rat liver cells after hepatocarcinogen treatment facilitates cell proliferation and slowing down of DNA damage response in GST-P-expressing preneoplastic lesions.
PMID 28899750
Abstract
We previously found downregulation of ubiquitin-conjugating enzyme E2E 2 (UBE2E2) in GST-P-positive