EMAIL THIS PAGE TO A FRIEND
International journal of molecular medicine
MicroRNA-132 induces temozolomide resistance and promotes the formation of cancer stem cell phenotypes by targeting tumor suppressor candidate 3 in glioblastoma.
PMID 28901390
Abstract
The prognosis of patients suffering from glioblastoma [also referred to as glioblastoma multiforme (GBM)] is dismal despite multimodal therapy. Chemotherapy with temozolomide may suppress tumor growth for a certain period of time (a few months); however, invariable tumor recurrence suggests that glioblastoma initiating cells (GICs) render these tumors persistant. Thus, the understanding of the molecular mechanisms of action of GICs as regards their role in the progression of GBM is important as such knowledge will be helpful in the discovery of novel drug targets, as well as in the design of novel therapeutic strategies for more effective treatment of the disease. In this study, we found that tumor suppressor candidate 3 (TUSC3) was downregulated in temozolomide-resistant U87MG cells (U87MG-res cells) and its restoration sensitized U87MG-res cells to temozolomide. TUSC3 was able to inhibit the formation of GIC phenotypes in the U87MG-res cells. The overexpression of microRNA (miR)‑132 inhibited TUSC3 protein expression in the U87MG cells. However, its overexpression did not degrade TUSC3 mRNA expression in the cells. miR‑132 was upregulated in the U87MG-res cells and its overexpression induced temozolomide resistance and the formation of cancer stem cell phenotypes in the U87MG cells. Thus, our data indicate that miR-132 induces temozolomide resistance and promotes the formation of cancer stem cell phenotypes by targeting TUSC3 in glioblastoma.