EMAIL THIS PAGE TO A FRIEND
Sensors (Basel, Switzerland)
Non-Destructive Evaluation of Historical Paper Based on pH Estimation from VOC Emissions.
PMID 28903284
Abstract
Volatile organic compounds (VOCs) emitted from materials during degradationcan be a valuable source of information. In this work, the emissions of furfural and aceticacid from cellulose were studied using solid-phase micro-extraction (SPME) incombination with gas chromatography-mass spectrometry. Two sampling techniques wereemployed: static headspace sampling using SPME for 1 h at 40