EMAIL THIS PAGE TO A FRIEND
Oncotarget
The G-protein-coupled bile acid receptor Gpbar1 (TGR5) protects against renal inflammation and renal cancer cell proliferation and migration through antagonizing NF-κB and STAT3 signaling pathways.
PMID 28903349
Abstract
Gpbar1 (TGR5), a G-protein-coupled bile acid membrane receptor, is well known for its roles in regulation of glucose metabolism and energy homeostasis. In the current work, we found that TGR5 activation by its ligand suppressed lipopolysaccharide (LPS)-induced proinflammatory gene expression in wild-type (WT) but not TGR5
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0330
S3I-201, ≥97% (HPLC)
C16H15NO7S