EMAIL THIS PAGE TO A FRIEND
Oncotarget
Extracorporeal shock wave treatment attenuated left ventricular dysfunction and remodeling in mini-pig with cardiorenal syndrome.
PMID 28903379
Abstract
This study tested the hypothesis that extracorporeal shock wave (ECSW) treatment can improve ischemia-induced left ventricular (LV) dysfunction in mini-pig with co-existing chronic kidney disease (CKD). LV ischemia in mini-pigs was induced by applying an ameroid constrictor over mid-left anterior descending artery (LAD), while model of CKD was established by right nephrectomy with partial ligation of left renal arterioles 2 weeks before LAD constriction. Thirty mini-pigs were randomly divided into group 1 (sham-control), group 2 (LV-ischemia), group 3 (LV-ischemia + CKD), Group 4 [LV-ischemia + ECSW (applied 1200 shots at 0.1 mJ/m