EMAIL THIS PAGE TO A FRIEND
Oncotarget
Cancer-associated mutations in the canonical cleavage site do not influence CD99 shedding by the metalloprotease meprin β but alter cell migration
PMID 28903388
Abstract
Transendothelial cell migration (TEM) is crucial for inflammation and metastasis. The adhesion molecule CD99 was shown to be important for correct immune cell extravasation and is highly expressed on certain cancer cells. Recently, we demonstrated that ectodomain shedding of CD99 by the metalloprotease meprin β promotes TEM
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU052611 MISSION® esiRNA, esiRNA human CD99 (esiRNA1)