EMAIL THIS PAGE TO A FRIEND
The Journal of toxicological sciences
Usefulness of urinary biomarkers for nephrotoxicity in cynomolgus monkeys treated with gentamicin, cisplatin, and puromycin aminonucleoside.
PMID 28904298
Abstract
The objective of this study was to investigate the availability of novel urinary biomarkers (BMs) such as total protein, albumin, β