EMAIL THIS PAGE TO A FRIEND
Cell host & microbe
Epitranscriptomic Enhancement of Influenza A Virus Gene Expression and Replication.
PMID 28910636
Abstract
Many viral RNAs are modified by methylation of the N