EMAIL THIS PAGE TO A FRIEND
Scientific reports
Synthesis of flower-like magnetite nanoassembly: Application in the efficient reduction of nitroarenes.
PMID 28912493
Abstract
A facile approach for the synthesis of magnetite microspheres with flower-like morphology is reported that proceeds via the reduction of iron(III) oxide under a hydrogen atmosphere. The ensuing magnetic catalyst is well characterized by XRD, FE-SEM, TEM, N