EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Inhibition of plant acetyl-coenzyme A carboxylase by the herbicides sethoxydim and haloxyfop.
PMID 2891354
Abstract
Incorporation of [14C]acetate or [14C]pyruvate into fatty acids in isolated corn seedling chloroplasts was inhibited 90% or greater by 10 microM sethoxydim or 1 microM haloxyfop. At these concentrations, neither sethoxydim nor haloxyfop inhibited [14C]acetate incorporation into fatty acids in isolated pea chloroplasts. Sethoxydim (10 microM) and haloxyfop (1 microM) did not inhibit incorporation of [14C]malonyl-CoA into fatty acids in cell free extracts from corn tissue cultures. Acetyl coenzyme A carboxylase (EC 6.4.1.2) from corn seedling chloroplasts was inhibited by both sethoxydim and haloxyfop, with I50 values of 2.9 and 0.5 microM, respectively. This enzyme in pea was not inhibited by 10 microM sethoxydim or 1 microM haloxyfop.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45817
Haloxyfop, PESTANAL®, analytical standard
C15H11ClF3NO4
36795
Sethoxydim, PESTANAL®, analytical standard
C17H29NO3S