EMAIL THIS PAGE TO A FRIEND
Oncotarget
Hexokinase 2 promotes tumor growth and metastasis by regulating lactate production in pancreatic cancer.
PMID 28915575
Abstract
Pancreatic ductal adenocarcinoma (PDAC) is a
Related Materials