EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
A hydrogen peroxide sensor based on TNM functionalized reduced graphene oxide grafted with highly monodisperse Pd nanoparticles.
PMID 28915946
Abstract
Addressed herein, we report the synthesis and characterization of a tert-nonyl mercaptan (TNM) functionalized reduced graphene oxide (rGO) supported palladium (Pd) nanoparticles (NPs) (Pd/TNM@rGO) as electrochemical sensor. The highly monodisperse Pd/TNM@rGO nanocomposite was applied for electrochemical determination of hydrogen peroxide (H