EMAIL THIS PAGE TO A FRIEND
Molecular therapy. Nucleic acids
Activation of AMPK Attenuated Cardiac Fibrosis by Inhibiting CDK2 via p21/p27 and miR-29 Family Pathways in Rats.
PMID 28918029
Abstract
Cardiac fibrosis is pathological damage associated with nearly all forms of heart disease. AMP-activated protein kinase (AMPK) is an evolutionary conserved energy-sensing enzyme. Emerging evidences indicate that AMPK plays an important role in cardiac fibrosis and cell proliferation. However, less is known about the detailed mechanism of AMPK activation on cardiac fibrosis. In this study, we found the AMPK activation improved the impaired cardiac function of cardiac fibrosis rats and decreased interstitial fibrosis. Further results indicated AMPK activation promoted p21 and p27 and inhibited CDK2 and cyclin E protein expressions both in vivo and in vitro. Moreover, AMPK activation repressed downstream transcription factor hepatocyte nuclear factor 4 alpha (HNF-4α) expression and decreased the binding of HNF-4α to TGF-β