EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Increased infiltration and tolerised antigen-specific CD8
PMID 28919997
Abstract
Human cytomegalovirus (HCMV) antigens in glioblastoma (GBM) present opportunities for personalised immunotherapy. However, their presence in GBM tissue is still under debate, and evidence of their impact on functional immune responses and prognosis is sparse. Here, we investigated the presence of pp65 (UL83) and immediate early 1 (IE-1) HCMV antigens in a cohort of Norwegian GBM patients (