EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Algoriphagus marisflavi sp. nov., isolated from water of an estuary environment.
PMID 28920831
Abstract
A Gram-stain-negative, non-motile, aerobic and rod-shaped or ovoid bacterial strain, designated KEM-106