EMAIL THIS PAGE TO A FRIEND
Nature cell biology
p38
PMID 28920954
Abstract
Receptor-interacting protein kinase-1 (RIPK1), a master regulator of cell fate decisions, was identified as a direct substrate of MAPKAP kinase-2 (MK2) by phosphoproteomic screens using LPS-treated macrophages and stress-stimulated embryonic fibroblasts. p38
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1452
TPCA-1, ≥95% (HPLC)
C12H10FN3O2S